Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-CAMK1G Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-1809R-CY5
BOSSBS-1809R-CY5EA 900 EUR InStock
BOSSBS-1809R-CY5
Anti-CAMK1G Rabbit Polyclonal Antibody (Cy5®)
Antikörper
CaMKI gamma is a Calcium/calmodulin-dependent protein kinase which belongs to a proposed calcium-triggered signaling cascade. In vitro it phosphorylates transcription factor CREB1. It has been proposed that CaMKI gamma is involved in dendritic outgrowth mediated by CaM-dependant protein kinase kinase.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CAMK1G
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge