Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-PAI2 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-1864R-A350
BOSSBS-1864R-A350EA 893 EUR InStock
BOSSBS-1864R-A350
Anti-PAI2 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
SerpinB2 is a serine proteinase inhibitor of the ovalbumin like B clade of serpins. It was first discovered in the placenta, and given the name PAI-2 because of the ability to inhibit urokinase plasminogen activator (u-PA) at low micromolar efficiency. SerpinB2 also inhibits tissue plasminogen activator (t-PA), but with micromolar efficiency, and PAI-1 is much more efficient that SerpinB2 on both forms of plasminogen activator. The structure of PAI-2 is not terribly similar to PAI-1, however, which is an E clade serpin. SerpinB2 is made by many cell types, and is found intracellularly as an unglycosylated kDa protein, and secreted as a 60 kDa protein. SerpinB2 is found in saliva, secreted by gingival fibroblasts, and in the skin. SerpinB2 levels are elevated in serum during pregnancy, and in leukemia, breast cancer and ovarian cancer, although it was lowered in some cancers.A shorter SerpinB2 isoform of 382 amino acids, has been reported, with a predicted mass of 43.1 kDa and a pI of 5.69. The shorter form has a deletion just after the start of the mature protein, but it is unclear what the relative production and distribution is for the shorter sequence.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PAI2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge