Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-Hepatitis B Virus X Protein Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-2147R-A680
BOSSBS-2147R-A680EA 890 EUR InStock
BOSSBS-2147R-A680
Anti-Hepatitis B Virus X Protein Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Multifunctional protein that may modulate protein degradation pathways, apoptosis, transcription, signal transduction, cell cycle progress, and genetic stability by directly or indirectly interacting with hosts factors. Does not seem to be essential for HBV infection. May be directly involved in development of cirrhosis and liver cancer (hepatocellular carcinoma). Most of cytosolic activities involve modulation of cytosolic calcium. The effect on apoptosis is controversial depending on the cell types in which the studies have been conducted. By binding to human DDB1, may affect cell viability and stimulate genome replication. May induce apoptosis by localizing in mitochondria and causing loss of mitochondrial membrane potential. May also modulate apoptosis by binding human CFLAR, a key regulator of the death-inducing signaling complex (DISC). Moderately stimulates transcription of many different viral and cellular transcription elements. Promoters and enhancers stimulated by HBx contain DNA binding sites for NF-kappa-B, AP-1, AP-2, c-EBP, ATF/CREB, or the calcium-activated factor NF-AT. May bind bZIP transcription factors like CREB1 (By similarity).

Type: Primary
Antigen: Hepatitis B virus X protein
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 10-60/210
Host: Rabbit
Isotype: IgG
Reactivity: Virus, Others

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge