Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-URGCP Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-2172R-CY5.5
BOSSBS-2172R-CY5.5EA 900 EUR InStock
BOSSBS-2172R-CY5.5
Anti-URGCP Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
URG4 may be involved in cell cycle progression through the regulation of cyclin D1 expression. It may participate in the development of hepatocellular carcinoma (HCC) by promoting hepatocellular growth and survival. URG4 may play an important role in development of gastric cancer.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: URGCP
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge