Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-KLF5 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-2385R-A488
BOSSBS-2385R-A488EA 901 EUR InStock
BOSSBS-2385R-A488
Anti-KLF5 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
The kinesin motor proteins include at least two forms of conventional kinesin encoded by different genes and designated as ubiquitous kinesin, which is expressed in all cells and tissues, or neuronal kinesin, which is expressed exclusively in neural cells. Kinesin is a microtubule associated protein comprised of three different structural domains. A considerable globular N-terminal domain regulates the hydrolysis of ATP and also microtubule binding while central coiled-coil domains promote heavy chain dimerization. Lastly, small globular C-terminal domains interact with kinesin light chains, membranous organelles and vesicles. Expression of ubiquitous kinesin heavy chain, also designated UKHC, is found subcellularly in areas of heavy vesicular trafficking such as the microtubule pathways of neural cells and also the Golgi of non-neural cell types.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: KLF5
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge