Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-MAPK3 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-3016R-CY3
BOSSBS-3016R-CY3EA 929 EUR InStock
BOSSBS-3016R-CY3
Anti-MAPK3 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
p44/42 MAP Kinase(Thr202); ERK (extracellular signal regulated kinase), also known as MAPK (mitogen activated protein kinase) has two closely related isoforms of 44 kDa and 42 kDa, respectively. These kinases belong to a family of serine/threonine kinases that are activated upon treatment of cells with a large variety of stimuli including mitogens, hormones, growth factors, cytokines, and bioactive peptides. Cell stimulation induces the activation of a signaling cascade, the downstream effects of which have been linked to the regulation of cell growth and differentiation as well as the cytoskeleton. ERK1 and ERK2 are phosphorylated within the activation loop on both a Threonine and a Tyrosine residue (within a Thr-Glu-Tyr motif) by MEKs (MAPK/ERK kinases), thereby greatly elevating the activity of ERK1&2.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: MAPK3
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Bovine

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge