Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-TSC2 SER939 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-3442R-FITC
BOSSBS-3442R-FITCEA 929 EUR InStock
BOSSBS-3442R-FITC
Anti-TSC2 SER939 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
Tuberin, or TSC2 (Tuberous sclerosis complex), is implicated as a tumor suppressor. It may function in vesicular transport, and may also play a role in the regulation of cell growth arrest and in the regulation of transcription mediated by steroid receptors. Interaction between hamartin (TSC1) and tuberin may facilitate vesicular docking. It specifically stimulates the intrinsic GTPase activity of the Ras related protein RAP1A and RAB5, suggesting a possible mechanism for its role in regulating cellular growth. Mutations in tuberin lead to constitutive activation of RAP1A in tumors. At least three isoforms of Tuberin exist.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Tuberin/TSC2
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge