Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-CTR9 Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-3697R-CY5.5
BOSSBS-3697R-CY5.5EA 900 EUR InStock
BOSSBS-3697R-CY5.5
Anti-CTR9 Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
CTR9 is a component of the PAF (polymerase-associated factor) complex which interacts with histone-modifying enzymes and RNA polymerase II and plays a role in a number of transcription-related processes. Recent studies show that CTR9 can regulate IL-6-responsive gene transcription by influencing histone methylation.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CTR9
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge