Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-SCAP Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-3862R-FITC
BOSSBS-3862R-FITCEA 883 EUR InStock
BOSSBS-3862R-FITC
Anti-SCAP Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
SCAP is an escort protein required for cholestrol and lipid homeostasis. Cholesterol homeostasis in animal cells is achieved by regulated cleavage of SREBPs, membrane-bound transcription factors. SCAP forms a complex with SREBPs in order to release the active domains of SREBPs.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SCAP
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge