Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-ADCY10 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-3916R-CY3
BOSSBS-3916R-CY3EA 900 EUR InStock
BOSSBS-3916R-CY3
Anti-ADCY10 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
Soluble adenylyl cyclase that has a critical role in mammalian spermatogenesis. Produces the cAMP which mediates in part the cAMP-responsive nuclear factors indispensable for maturation of sperm in the epididymis. Induces capacitation, the maturational process that sperm undergo prior to fertilization. May be the bicarbonate sensor. Involved in ciliary beat regulation.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ADCY10
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge