Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-CHFR Rabbit Polyclonal Antibody

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-4272R
BOSSBS-4272REA 725 EUR InStock
BOSSBS-4272R
Anti-CHFR Rabbit Polyclonal Antibody
Antikörper
A number of human cancers are sensitive to mitotic stress, implying checkpoint defects. Many proteins that contain forkhead-associated (FHA) domains are cell cycle checkpoints and CHFR is one such protein. It has a FHA and ring finger domain within its N terminus. Within its C terminus, CHFR contains a cysteine-rich region that did not display significant similarity to any protein in the GenBank database, but is highly conserved between humans and mice. It has been concluded that CHFR defines a checkpoint that delays entry into metaphase in response to mitotic stress. It has been found CpG methylation-dependent silencing of CHFR expression occurs in 45% of cancer cell lines, 40% of primary colorectal cancers, 53% of colorectal adenomas, and 30% of primary head and neck cancers.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: CHFR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge