Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-NRIP Rabbit Polyclonal Antibody

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-6210R
BOSSBS-6210REA 725 EUR InStock
BOSSBS-6210R
Anti-NRIP Rabbit Polyclonal Antibody
Antikörper
WD-repeat proteins are a large family of eukaryotic proteins coordinating multi-protein complex assemblies. Their role has been implicated in multiple cellular processes including signal transduction, transcriptional regulation, cell cycle control and apoptosis. NRIP is a novel 860a.a nuclear protein consisting of seven conserved WD40 domains and one NLS motif. It binds to androgen and glucocorticoid receptors and up-regulates their transcriptional activity, thereby functioning as a nuclear receptor co-activator. Role of NRIP has been implicated in cell growth and also in cervical and prostrate cancer, thus indicating a potential therapeutic intervention. Northern Blot analysis detects a high expression of NRIP in skeletal muscle and testis and low expression in heart, prostrate and adrenal gland.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: NRIP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge