Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-MYOT Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-6479R-A647
BOSSBS-6479R-A647EA 893 EUR InStock
BOSSBS-6479R-A647
Anti-MYOT Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
Myotilin, a sarcomeric protein that is encoded by the gene mapping to human chromosome 5q31, binds to a-actinin and is localized in the Z-line of myofibrils. Myotilin is expressed in skeletal and cardiac muscle, and it co-localizes with a-actinin in the sarcomeric I-bands where it directly interacts with a-actinin. Defects in the myotilin gene are reported to cause a form of autosomal dominant limb-girdle muscular dystrophy (LGMD). Symptoms of adult onset LGMD are progressive weakness of the hip and shoulder girdles as well as a distinctive dysarthric pattern of speech. The muscle of affected individuals with LGMD shows degeneration of myofibers, variations in fiber size, fiber splitting, centrally located myonuclei and an enhanced number of autophagic vesicles.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: MYOT
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge