Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-LRDD Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-7615R-A647
BOSSBS-7615R-A647EA 893 EUR InStock
BOSSBS-7615R-A647
Anti-LRDD Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
The death domain (DD) containing protein PIDD is a p53 target gene in an erythroleukemia cell line that undergoes G1 phase arrest and subsequent apoptosis after p53 expression. Independently, PIDD was also described as a DD-containing protein with unknown function. The N-terminal region of PIDD contains seven leucine-rich repeats (LRRs), a protein interaction motif found in various proteins with diverse functions, followed by two ZU-5 domains and a C-terminal DD. PIDD forms a complex with caspase-2 and the adaptor protein RAIDD. Increased PIDD expression results in spontaneous activation of caspase-2 and sensitization to apoptosis by genotoxic stimuli, via interaction with caspase-2 and CRADD/RAIDD. PIDD also promotes apoptosis downstream of p53 as component of the DNA damage/stress response pathway that connects p53/TP53 to apoptosis. PIDD has also been shown to interact with NEMO/IKBKG and RIP1 and enhance sumoylation and ubiquitination of NEMO/IKBKG, an important component for activation of the transcription factor NF-kappa-B.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: LRDD
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge