Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-DNAJA3 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-7645R-A488
BOSSBS-7645R-A488EA 893 EUR InStock
BOSSBS-7645R-A488
Anti-DNAJA3 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
TID1 is a human homolog of the Drosophila tumor suppressor lethal tumerous imaginal discs and encodes two mitochondrial matrix localized splice variants of human Tid1 designated hTid1S and hTid1L. These proteins are the conserved members of the DnaJ family of proteins which act as cochaperons for mitochondrial Hsp70. They contain a conserved tetrahedrical J domain which binds to Hsp70 chaperones and activates their ATPase activity. Expression of hTid1L increases apoptosis induced by DNA damaging agents as mitomycin C and TNF alpha. A J domain mutant of hTid1L can dominantly suppress apoptosis and in sharp contrast the J domain mutant of hTid1S increases apoptosis. Expression of hTid1S and hTid1L affects cytochrome c release from the mitochondria and caspase 3 activation, while activation of caspase 8 is unaffected. It is strongly suggested that these two splice variants exert their anti and pro apoptotic effects through discrete substrates and activities. Hence the relative abundance of these proteins or their substrates may allow the mitochondria to dampen or enhance the apoptotic signals.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: DNAJA3
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge