Anti-DNASE1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Lieferant: Bioss
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
BS-7651R-HRP
BOSSBS-7651R-HRPEA 900 EUR InStock
BOSSBS-7651R-HRP
Anti-DNASE1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Deoxyribonuclease I gene is approximately 3.2 kb long with 9 exons separated by 8 introns. In the form of a bovine pancreatic enzyme preparation, it occupies an important place in the history of protein chemistry and enzymology: it was the first enzyme to be recognized as specific for DNA; it was the first DNase to be crystallized; and it was the first DNase for which a specific protein inhibitor was characterized. DNase I is a Ca2+ and Mg2+ dependant endonuclease. DNase I is synthesized in the pancreas and stored in zymogen granules. It has been used to reduce the viscosity of cysticMore Product Information
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge
Diese Webseite verwendet Cookies, eigene oder von Drittanbieter, zur Sammlung statistischer Informationen, um Inhalte und Anzeigen für Sie zu personalisieren. Wenn Sie diese Seite weiterhin verwenden, akzeptieren Sie die Cookie-Nutzung.
Weitere Informationen finden Sie in unseren Datenschutzbestimmungen..