Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-HCFC1 Rabbit Polyclonal Antibody

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-7803R
BOSSBS-7803REA 725 EUR InStock
BOSSBS-7803R
Anti-HCFC1 Rabbit Polyclonal Antibody
Antikörper
Involved in control of the cell cycle. Also antagonizestransactivation by ZBTB17 and GABP2; represses ZBTB17 activation ofthe p15(INK4b) promoter and inhibits its ability to recruit p300.Coactivator for EGR2 and GABP2. Tethers the chromatin modifyingSet1/Ash2 histone H3 'Lys-4' methyltransferase (H3K4me) and Sin3histone deacetylase (HDAC) complexes (involved in the activationand repression of transcription, respectively) together. Componentof a THAP1/THAP3-HCFC1-OGT complex that is required for theregulation of the transcriptional activity of RRM1. As part of theNSL complex it may be involved in acetylation of nucleosomalhistone H4 on several lysine residues. In case of human herpessimplex virus (HSV) infection, HCFC1 forms a multiprotein-DNAcomplex with the viral transactivator protein VP16 and POU2F1thereby enabling the transcription of the viral immediate earlygenes.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: HCF-1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge