Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-ARRDC4 Rabbit Polyclonal Antibody (HRP)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-8078R-HRP
BOSSBS-8078R-HRPEA 1350 EUR InStock
BOSSBS-8078R-HRP
Anti-ARRDC4 Rabbit Polyclonal Antibody (HRP)
Antikörper
ARRDC4 belongs to the arrestin family. The arrestins are a family of proteins that are important for regulating signal transduction within cells. Arrestins are part of a conserved two step mechanism for regulating the activity of G-protein coupled receptors (GPCRs). In response to a stimulus, GPCRs activate a heterotrimeric G protein. In order to turn off this response, or adapt to a constant stimulus, activated receptors need to be silenced. The first step is phosphorylation by a class of serine/threonine kinases called G protein coupled receptor kinases (GRKs). This phosphorylation specifically marks the activated receptor for arrestin binding. Once arrestin is bound to the receptor it is unable to signal further. Recent research continues to expand the known actions of arrestins, which can bind to other classes of receptors and can directly activate signaling pathways on their own. Different arrestins (visual arrestin (or Arrestin 1), beta-arrestin 1 (or Arrestin 2) and beta-arrestin 2 (or Arrestin 3) can reduce the activity of their target GPCRs in several different ways.

Type: Primary
Antigen: ARRDC4
Clonality: Polyclonal
Clone:
Conjugation: HRP
Public Immunogen Range: 201-330/418
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge