Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-DIRC2 Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-8278R-CY5
BOSSBS-8278R-CY5EA 900 EUR InStock
BOSSBS-8278R-CY5
Anti-DIRC2 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
Belonging to the major facilitator superfamily, DIRC2 (disrupted in renal carcinoma protein 2) is a 478 amino acid multi-pass membrane protein that is primarily expressed in kidney proximal tubular cells. The genes encoding DIRC2 and DIRC3 are located at a translocation breakpoint which occurs frequently in individuals affected by familial renal cell carcinoma. Fusion transcripts that result from these translocations may affect normal protein function. This evidence suggests that, due to its chromosomal location, deregulation of the DIRC2 gene may cause haploinsufficiency and therefore result in the onset of tumor growth. There are two isoforms of DIRC2 which are produced as a result of alternative splicing events.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: DIRC2
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge