Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-DPY19L1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-8289R-A680
BOSSBS-8289R-A680EA 893 EUR InStock
BOSSBS-8289R-A680
Anti-DPY19L1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Dpy-19 (dumpy-19), is a 683 amino acid C. elegans protein that is required to orient the neuroblasts QL and QR correctly on the anterior/posterior axis. Dpy-19 is expressed highly in dorsal hyp7 cells, ventral P cells and lateral V cells, and dorsal and ventral body muscle cells. DPY19L1 (Dpy-19-like protein 1), also known as KIAA0877, is a 675 amino acid multi-pass membrane protein that belongs to the Dpy-19 family. DPY19L1 is expressed as two isoforms produced by alternative splicing and is encoded by a gene mapping to human chromosome 7, which encodes over 1,000 genes and makes up about 5% of the human genome. Diseases associated with chromosome 7 include Osteogenesis imperfecta, Pendred syndrome, Lissencephaly, Citrullinemia and Shwachman-Diamond syndrome. The deletion of a portion of the q arm of chromosome 7 is associated with Williams-Beuren syndrome, a condition characterised by mild mental retardation, an unusual comfort and friendliness with strangers and an elfin appearance. Deletions of portions of the q arm of chromosome 7 are also seen in a number of myeloid disorders including cases of acute myelogenous leukemia and myelodysplasia.

Type: Primary
Antigen: DPY19L1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 571-675/675
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge