Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-UBE2N Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-8351R-A488
BOSSBS-8351R-A488EA 893 EUR InStock
BOSSBS-8351R-A488
Anti-UBE2N Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
Ubiquitination is an important mechanism through which three classes of enzymes act in concert to target short-lived or abnormal proteins for destruction. The three classes of enzymes involved in ubiquitination are the ubiquitin-activating enzymes (E1s), the ubiquitin-conjugating enzymes (E2s) and the ubiquitin-protein ligases (E3s). UBC13, also known as UBE2N or BLU, is a 152 amino acid member of the E2 ubiquitin-conjugating enzyme family. Existing as a heterodimer with Mms2 (also known as UBE2V2), UBC13 catalyzes the ATP-dependent synthesis of non-canonical polyubiquitin chains, a process that does not lead to proteasomal degradation. Additionally, UBC13 mediates the transcription of several target genes and is thought to play a role in cell cycle progression, cellular differentiation and DNA repair mechanisms that ensure cell survival after DNA damage.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: UBE2N
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge