Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-BPIFB2 Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-8423R-CY7
BOSSBS-8423R-CY7EA 900 EUR InStock
BOSSBS-8423R-CY7
Anti-BPIFB2 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
Members of the bactericidal/permeability-increasing protein family have antimicrobial properties and bind lipophilic substances, therefore targeting gram-negative bacteria. The bactericidal permeability increasing protein (BPI) is an antibacterial and endotoxin-neutralizing molecule that is abundant in the granules of polymorphonuclear leukocytes (neutrophil granules). Sharing structural and sequence homologies with BPI, BPIL1 (bactericidal/permeability-increasing protein-like 1) is a 458 amino acid secreted protein that contains the family’s common conserved feature of two cysteine residues that are critical for protein function. While BPIL1 is primarily expressed at low levels in tonsil tissue, it has been found to be upregulated in hypertrophic tonsils, suggesting that it may play a role in the pathogenesis of inflamed disease tissue.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: BPIFB2
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge