Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-RANBP7 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-8590R-A555
BOSSBS-8590R-A555EA 893 EUR InStock
BOSSBS-8590R-A555
Anti-RANBP7 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
Importin 7 is a 1,038 amino acid protein encoded by the human gene IPO7. Importin-7 belongs to the importin b family and contains one importin N-terminal domain. Importin-7 functions in nuclear protein import, either by acting as an autonomous nuclear transport receptor or as an adapter-like protein in association with the Importin b subunit KPNB1. Acting autonomously, Importin-7 is thought to serve itself as receptor for nuclear localization signals (NLS) and to promote translocation of import substrates through the nuclear pore complex (NPC) by an energy requiring, Ran-dependent mechanism. At the nucleoplasmic side of the NPC, Ran binds to Importin-7, the Importin-7/substrate complex dissociates and Importin-7 is re-exported from the nucleus to the cytoplasm where GTP hydrolysis releases Ran. Importin-7 is a nuclear protein that is expressed in most tissues.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: RANBP7
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge