Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-IQCG Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9022R-A750
BOSSBS-9022R-A750EA 893 EUR InStock
BOSSBS-9022R-A750
Anti-IQCG Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
IQCG is a 443 amino acid protein containing one IQ domain. Widely distributed in nature, the IQ domain forms an amphiphilic seven-turn -helix capable of binding calmodulin in a Ca2+-independent manner. The level of intracellular calcium is tightly regulated in all eukaryotic cells. A modest increase in this level can result in a myriad of physiological responses, most of which are mediated by calmodulin (CaM), the universal calcium sensor. In acute T-lymphoid/myeloid leukemia, IQCG forms a complex with Nup98, an O-linked glycoprotein and a component of the nuclear pore complex. NUP98-IQCG complex bind co-activators and/or co-repressors, which suggest a role in transcriptional regulation.Nup98-IQCG complex inhibits 32Dcl3 cell apoptosis induced by Arabinofuranosylcytosine (Ara-C) and partially blocks granulocyte differentiation induced by G-CSF. IQCG exists as two isoforms due to alternatively splicing events.

Type: Primary
Antigen: IQCG
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 301-400/443
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge