Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-CDK5RAP1 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9075R-CY3
BOSSBS-9075R-CY3EA 900 EUR InStock
BOSSBS-9075R-CY3
Anti-CDK5RAP1 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
Cyclin dependent kinase 5 (Cdk5) is a key regulator of cell cycle progression in neuronal differentiation that physically associates with and is activated by the neuron-specific protein p35. CDK5RAP1 (Cdk5 regulatory subunit-associated protein 1), also known as Cdk5 activator-binding protein C42, is a 601 amino acid protein that specifically inhibits Cdk5 activation by p35 through formation of a dimer that inhibits kinase activity. CDK5RAP1 contains one TRAM domain, which is thought to bind tRNA and deliver the RNA-modifying enzymatic domain to its target. There are 4 named isoforms of CDK5RAP1 that are produced as a result of alternative splicing events and are expressed at high levels in heart and skeletal muscle.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CDK5RAP1
Clonality: Polyclonal
Clone: 0
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge