Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-VGLL4 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9185R-HRP
BOSSBS-9185R-HRPEA 900 EUR InStock
BOSSBS-9185R-HRP
Anti-VGLL4 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
The transcriptional enhancer factor-1 (TEF-1) family of transcription factors regulate tissue-specific gene expression in muscle and placenta. The mechanism whereby TEF-1 confers tissue specificity depends largely on the interaction of TEF-1 with tissue-specific cofactors. Transcription cofactor Vgl-4 (vestigial-like protein 4) is a 290 amino acid nuclear protein that interacts with TEF-1 and MEF-2. Vgl-4 is the only member of the vestigial-like family that is expressed in heart. Overexpression of Vgl-4 in cardiac myocytes interferes with basal expression and ?-adrenergic receptor-dependent activation of a TEF-1 dependent skeletal ?actin promoter. This suggests that Vgl-4 counteracts ?-adrenergic activation of gene expression in cardiomyocytes. There are two isoforms of Vgl-4 that are produced as a result of alternative splicing events.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: VGLL4
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge