Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-IGSF11 Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9201R-CY5.5
BOSSBS-9201R-CY5.5EA 900 EUR InStock
BOSSBS-9201R-CY5.5
Anti-IGSF11 Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
IGSF11 is also known as BTIGSF (brain and testis-specific immunoglobulin superfamily protein) or VSIG3 (V-set and immunoglobulin domain-containing protein 3) and is a 431 amino acid protein that is expressed as three isoforms. IGSF11 is highly expressed in testis and ovary and is also expressed in brain, kidney and skeletal muscle, localized to the cellular membrane as a single-pass membrane protein. IGSF11 is an immunoglobulin with V-type and C2-type domains that function in molecular recognition. When IGSF11 is in the trans position, it plays an important role in cell-cell adhesion via both homophilic and heterophilic interactions with other molecules. These cell–cell interactions are also thought to be important for neuronal cell interactions, such as neuron–neuron or neuron–glia interactions, which are important for the development and function of the central nervous system. In addition, IGSF11 might also be involved interactions between Sertoli cells and spermatocytes, which are important associations during spermatogenesis. The IGSF11 gene is commonly upregulated in gastric cancer and IGSF11 is highly expressed in many types of human tumors, indicating that it may be useful as a target for immunotherapy.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: IGSF11
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge