Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-SASS6 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9301R-A680
BOSSBS-9301R-A680EA 893 EUR InStock
BOSSBS-9301R-A680
Anti-SASS6 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
SAS-6 (spindle assembly abnormal protein 6 homolog, HsSAS-6) is a 657 amino acid protein encoded by the human gene SAS6. SAS-6 is a component of the centrosome that contains one PISA (present in SAS-6) domain. LK4, SAS-6, CPAP and other centriole related proteins are required at different stages of procentriole formation and were associated with different centriolar structures. SAS-6 associates only transiently with nascent procentrioles, whereas CEP135 and CPAP form a core structure within the proximal lumen of both parental and nascent centrioles. SAS-6 is necessary for procentriole formation in human cell lines and is localised asymmetrically next to the centriole at the onset of procentriole formation. SAS-6 levels oscillate during the cell cycle; it is degraded in mitosis starting at anaphase, and it accumulates again at the end of the following G1 phase. The anaphase-promoting complex targets SAS-6 for degradation by the 26S Proteasome, and a KEN box in the C-terminus of SAS-6 is necessary for its degradation. Increased SAS-6 levels promoted the formation of multiple procentrioles forming next to a single centriole.

Type: Primary
Antigen: SASS6
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 161-260/657
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge