Anti-LRCH3 Rabbit Polyclonal Antibody (Cy3®)
Lieferant: Bioss
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
BS-9329R-CY3
BOSSBS-9329R-CY3EA 900 EUR InStock
BOSSBS-9329R-CY3
Anti-LRCH3 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
Members of the leucine-rich repeat family includes LRCH1, LRCH2, LRCH3 and LRCH4. All family members contain one calponin-homology domain and nine leucine-rich repeats. The best characterized leucine-rich repeat family member is LRCH4, which is suggested to be involved in ligand binding in the brain, with expression observed primarily in the hippocampus. As a cell adhesion molecule and signal receptor, LRCH4 may play an important role in maintenance of hippocampus-dependent memories, with defects in the gene possibly contributing to a loss of long-term memory. The gene encoding LRCH3 maps to human chromosome 3, which spans 200 million base pairs and encodes between 1,100 and 1,500 genes. There are three isoforms of LRCH3 that are produced as a result of alternative splicing events.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: LRCH3
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge