Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-KChIP2 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9441R-A750
BOSSBS-9441R-A750EA 893 EUR InStock
BOSSBS-9441R-A750
Anti-KChIP2 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
The downstream regulatory element, DRE, acts as a location-dependent gene silencer. DREAM (for DRE-antagonist modulator) is a Ca²⁺ regulated transcriptional repressor that specifically binds to the DRE. DREAM regulates transcription of prodynorphin and c-Fos genes and shows 99% nucleotide homology to the Kv channel-interacting proteins (KChIPs). KChIP family members include KChIP1, which is expressed in brain, KChIP2, which is expressed in heart, brain, and lung, KChIP3 (also designated calsenilin), which is expressed in brain and testis and KChIP4.

Type: Primary
Antigen: KChIP2
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 1-100/270
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge