Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-JMJD7 Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9486R-CY5.5
BOSSBS-9486R-CY5.5EA 900 EUR InStock
BOSSBS-9486R-CY5.5
Anti-JMJD7 Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
A crucial regulator of chromatin dynamics and DNA transcription is the covalent modification and methylation of histones. Generally, methylation of certain lysine residues on Histone H3 and Histone H4 can be associated with transcriptionally active or inactive chromatin. This regulation has profound effects on the expression of genes and is part of an epigenetic memory network that determines cell fate. JMJD7 (Jumonji domain-containing protein 7) is a member of a family of JMJC domain-containing histone demethylases that are directly involved in removing methyl residues from distinct and unique lysine residues. These actions are implicated in gene expression and the regulation of cell senescence. JMJC domain-containing histone demethylases are also likely involved in development via their ability to regulate gene expression. JMJD7 contains one JMJC domain and was originally thought to be an alternatively spliced isoform of PLA2G4B.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: JMJD7
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge