Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-EPR1 Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9507R-CY5.5
BOSSBS-9507R-CY5.5EA 900 EUR InStock
BOSSBS-9507R-CY5.5
Anti-EPR1 Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
Cellular receptors for blood proteases regulate chemotaxis, extracellular proteolysis, and growth behavior of normal and malignant cells. Effector cell protease receptor-1 (EPR1) is a receptor for the coagulation protease factor Xa. EPR1 is characterized by a cysteine-rich extracellular module, a single membrane-spanning domain, and a serine-rich cytoplasmic tail featuring at least 15 potential phosphorylation sites. EPR1 also contains 2 N-linked glycosylation sites, 4 O-linked glycosylation sites, and a chondroitin sulfate attachment site, which may provide anchoring for carbohydrate chains, EPR1 transfectants bind to factor Xa in a specific and saturable manner, and in the absence of factor V/Va promote prothrombin activation in a factor Xa concentration-dependent reaction. Activated platelets and megakaryocytes express EPR1. Both EPR1 and membrane-bound factor Va are thought to be required to mediate factor Xa binding to the activated platelet to form a functional prothrombinase complex.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: EPR1
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge