Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-SNCA Rabbit Polyclonal Antibody

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9587R
BOSSBS-9587REA 725 EUR InStock
BOSSBS-9587R
Anti-SNCA Rabbit Polyclonal Antibody
Antikörper
The synuclein family members, including Alpha-synuclein (also designated NACP for non-Beta-Amyloid component) and Beta-synuclein, are predominantly expressed in the brain and are speculated to be involved in synaptic regulation and neuronal plasticity. Alpha-synuclein is localized to neuronal cell bodies and synapses. Alpha-synuclein was first identified as a component of Alzheimer’s disease amyloid plaques. Abnormal platelet function in Alzheimer’s disease has been demonstrated. During megakaryocytic differentiation Alpha-synuclein has been found to be upregulated, while Beta-synuclein is downregulated, indicating that coordinate expression of synucleins may be important during hematopoetic cell differentiation. A mutant form of Alpha-synuclein has been found in patients with early onset Parkinson’s disease.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: SNCA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge