Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-C20orf11 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9689R-A680
BOSSBS-9689R-A680EA 893 EUR InStock
BOSSBS-9689R-A680
Anti-C20orf11 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
TWA1 is a 228 amino acid nuclear protein that is highly conserved throughout evolution. TWA1 interacts with Ran BP-M, a protein that is implicated in a diverse array of cellular processes including DNA replication, entry into and exit from mitosis, and the transport of RNA and proteins through the nuclear pore complex. Together, Ran BP-M and TWA1 form a complex with cytoplasmic Muskelin, a mediator of cell spreading. It is thought that this complex plays a role in the Ran GTPase cycle and therefore a potential role in the cell cycle. TWA1 contains a LisH-CTLH motif which is usually found in proteins that are involved in nucleokinesis, chromosome segregation, cell migration and microtubule dynamics. The gene encoding TWA1 maps to human chromosome 20, which comprises approximately 2% of the human genome and contains nearly 63 million bases that encode over 600 genes.

Type: Primary
Antigen: C20orf11
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 1-100/228
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge