Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-C10ORF27 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9720R-HRP
BOSSBS-9720R-HRPEA 900 EUR InStock
BOSSBS-9720R-HRP
Anti-C10ORF27 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
C10orf27, also known as spatial, is a 351 amino acid cytoplasmic protein belonging to the spatial family. C10orf27 is suggested to play a role in spermatid differentiation. Existing as two alternatively spliced isoforms, C10orf27 is widely expressed in multiple tissues, including brain, thymus and testis. C10orf27 may be associated with multiple sclerosis (MS) susceptibility and pathogenesis. MS is an inflammatory disease that causes gradual destruction of myelin in the central nervous system. C10orf27 is encoded by a gene located on human chromosome 10, which contains over 800 genes and 135 million nucleotides, making up nearly 4.5% of the human genome. PTEN is an important tumor suppressor gene located on chromosome 10 and, when defective, causes a genetic predisposition to cancer development known as Cowden syndrome.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: C10ORF27
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge