Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-CHPT1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9725R-HRP
BOSSBS-9725R-HRPEA 900 EUR InStock
BOSSBS-9725R-HRP
Anti-CHPT1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
CHPT1, also known as AAPT1-like protein and Diacylglycerol cholinephosphotransferase 1, is a 406 amino acid multi-pass membrane protein that is localized to the golgi apparatus. By catalyzing the phosphatidylcholine biosynthesis from CDP-choline, it plays an essential role in the formation and maintenance of vesicular membranes. CHPT1 is most abundant in testis, as well as small intestine, heart, colon, spleen and prostate. Expression of CHPT1 is increased in cancerous breast cells as compared to normal breast cell lines and it has been determined that the CHPT1 gene exhibits mutations within the cancerous cells. Interestingly, exposure to mustard gas significantly decreases CHPT1 gene expression and activity, an event that may play an important role in the development of acute respiratory distress syndrome (ARDS). There are two isoforms of CHPT1 that are produced as a result of alternative splicing events.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: CHPT1
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge