Labels for thermal transfer printers, Type B-428, Brady
Lieferant: Brady
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
030108 030125 030133 030144 030147 030247
BRDY030108EA 573.05 EUR InStock
BRDY030108 BRDY030125 BRDY030133 BRDY030144 BRDY030147 BRDY030247
Labels for thermal transfer printers, Type B-428, Brady
Etiketten
Printable, Metallised polyester labels for thermal transfer printers.
  • Fuel/oill resistance
  • High heat resistance
  • Low temperature resistance
  • Solvent/chemical resistance
Zertifizierungen: This material is UL, CSA and AGA approved with its respective ribbon.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge