Workstation Automation Software Suite
Lieferant: Brady
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
149187
BRDY149187EA 832.99 EUR InStock
BRDY149187
Workstation Automation Software Suite
Software Labor-Software
Save time and reduce errors with data Automation. By automatically importing data onto a pre-defined label template and printing to a designated printer, this app allows label printing to seamlessly become a part of your daily processes. It runs in the background so you won't have to interrupt your work just to print a label.
Idel for asset tracking, barcode labelling, data and telecommunications labelling, electrical labelling, general identification, inventory and inspection labelling.
Operating System:
Windows® 10, Windows® 11, Windows® 8, Windows® 8.1, Windows® Server 2012, Windows® Server 2016, Windows® Server 2019, Windows® Server 2022.
Languages available: Bulgarian, Chinese (Simplified), Chinese (Traditional), Croatian, Czech, Danish, Dutch, English (US), Estonian, Finnish, French, German, Hungarian, Italian, Japanese, Korean, Norwegian (Bokmål), Polish, Portuguese, Romanian, Russian, Slovak, Slovenian, Spanish, Spanish (Mexico), Swedish, Turkish, Vietnamese.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge