SPC® Absorbents SitePoint Spill Ready Kit
Lieferant: Brady
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
309077
BRDY309077EA 712.29 EUR InStock
BRDY309077
SPC® Absorbents SitePoint Spill Ready Kit
Absorptionsmaterial
Spill kit.
  • Loaded SitePoint Mobile Unit with suitable products for cleaning up general spills
  • Features two, 200 mm wheels for mobility and anchor points for fixed installation
  • Fix items to backboard using self-tapping screws or bolts
  • Suitable for inside or outdoor use
Ideal to move spill control equipment near specific risks. Features two rubber wheels for easy trolley movement to wherever it is required. Perfect during maintenance activities, at construction sites and other temporary locations. Includes three anchor points on the base, allowing the unit to be bolted to the ground, to avoid trolley movement without permission.
Lieferumfang: Includes 1×SitePoint mobile unit, 1×scoop, 1×economy maintenance spill kit, 1×QuickWik, 1×floor stand and fixings.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge