Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-SMAD4 Rabbit Polyclonal Antibody

Lieferant: BosterBio

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

PB9397
BSBTPB9397EA 552 EUR InStock
BSBTPB9397
Anti-SMAD4 Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for Mothers against decapentaplegic homolog 4(SMAD4) detection. Tested with WB in Human;Mouse;Rat.

SMAD4 (Mothers Against Decapentaplegic Drosophila Homolog of 4), also known as MADH4 or DPC4, is a protein that in humans is encoded by the SMAD4 gene. It belongs to the Darfwin family of proteins that modulate members of the TGF? protein superfamily. Hahn et al. (1996) identified the SMAD4 gene on chromosome 18q21.1. Howe et al. (1998) identified the SMAD4 gene within a region on 18q21.1 defined by linkage analysis in kindred with juvenile polyposis syndrome. To test directly the hypothesis that the SMAD4 gene is a tumor suppressor that is critical for transmitting signals from transforming growth factor-beta and related ligands. SMAD4 plays a pivotal role in signal transduction of the transforming growth factor beta superfamily cytokines by mediating transcriptional activation of target genes.

Add 0.2ml of distilled water will yield a concentration of 500ug/ml.

Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg NaN3.

WB: The detection limit for Smad4 is approximately 0.1ng/lane under reducing conditions.
Tested Species: In-house tested species with positive results.
Other applications have not been tested.
Optimal dilutions should be determined by end users.

Type: Primary
Antigen: SMN1/2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge