Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-TGF-1 Chicken Polyclonal Antibody

Lieferant: Biosensis

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

C-1541-100
BSENC-1541-100EA 321 EUR InStock
BSENC-1541-100
Anti-TGF-1 Chicken Polyclonal Antibody
Antikörper
Transforming growth factor beta-1 (TGFB1) is a multi-functional cytokine with roles in proliferation, differentiation and other functions in many cell types. The secreted TGFB1 protein is cleaved into a latency-associated peptide (LAP) and a mature TGFB1 peptide (Ref: SWISS-Prot).

Antibody recognizes TGFB1 by western blot and ELISAs; works best with cell supernatants or fluids as TGFB1 is secreted. Other applications have not been tested.
Application Information:
WB and ELISA. Suggested dilution of 1:2,000-1:5,000. Biosensis recommends that the optimal working dilution should be determined by the end user.

Type: Primary
Antigen: TGF-1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Human TGFbeta-1 amino acids 372-386
Host: Chicken
Isotype: IgY
Reactivity: Human, Mouse, Rat, Pig

Order Now
 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge