Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-TAR DNA-binding Protein 43 Mouse Monoclonal Antibody [clone: 3H8]

Lieferant: Biosensis

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

M-1403-100
BSENM-1403-100EA 312 EUR InStock
BSENM-1403-100
Anti-TAR DNA-binding Protein 43 Mouse Monoclonal Antibody [clone: 3H8]
Antikörper
TAR DNA-binding protein 43 (TDP43) is a DNA and RNA-binding protein which regulates transcription and splicing (ref: SWISSPROT).

The specificity of this antibody has been confirmed by WB. This antibody detects ~43 kDa TDP43 protein on crude extract of mouse brain nuclear fraction.
Application Information:
Western Blotting (WB) and Immunocytochemistry (IC). A dilution of 1:1,000 - 1:5,000 is recommended for WB. A dilution of 1:500-1,000 is recommended for IC. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: TAR DNA-binding Protein 43
Clonality: Monoclonal
Clone: 3H8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human, Rodent

Order Now
 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge