Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-SAP6 Mouse Monoclonal Antibody [clone: X63] Saporin

Lieferant: Biosensis

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

M-1792-25-IT M-1792-50-IT M-1792-100-IT
BSENM-1792-25-ITEA 245 EUR InStock
BSENM-1792-25-IT BSENM-1792-50-IT BSENM-1792-100-IT
Anti-SAP6 Mouse Monoclonal Antibody [clone: X63] Saporin
Antikörper
X63-saporin is an antibody conjugate comprising of the non-specific monoclonal IgG1 antibody X63, chemically conjugated via a reducible disulfide bridge to the ribosome-inactivating protein saporin, purified from saponaria officinalis. Antibody clone X63 has no known binding ability, and thus can be used as negative control antibody. In combination with saporin, X63-saporin is a useful negative control for targeting IgG1-saporin conjugates such as MC192-saporin.

  • Non-reactive (negative control)

X63-saporin is an antibody conjugate comprising of the non-specific monoclonal IgG1 antibody X63, chemically conjugated via a reducible disulfide bridge to the ribosome-inactivating protein saporin, purified from saponaria officinalis. Antibody clone X63 has no known binding ability, and thus can be used as negative control antibody. In combination with saporin, X63-saporin is a useful negative control for targeting IgG1-saporin conjugates such as MC192-saporin.

Other applications: Antibody clone X63 has no known binding ability, and thus can be used as negative control antibody.

Achtung! Bitte beachten: Only qualified personnel should handle this product. X63 antibody has no known target receptor, however, standard laboratory protective measures should be followed, including wearing lab coat, gloves and safety glasses. Avoid contact with open wounds and practice good laboratory technique to avoid auto-exposure/auto-injection! Avoid contact of conjugate with reducing agents (eg., DTT, beta-mercaptoethanol) as this will destroy the antibody-saporin conjugate.

Type: Primary
Antigen:
Clonality: Monoclonal
Clone: X63
Conjugation: Saporin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge