Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-GFR alpha-3 Rabbit Polyclonal Antibody

Lieferant: Biosensis

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

R-068-100
BSENR-068-100EA 312 EUR InStock
BSENR-068-100
Anti-GFR alpha-3 Rabbit Polyclonal Antibody
Antikörper
FUNCTION: Receptor for the glial cell line-derived neurotrophic factor, artemin. Mediates the artemin-induced autophosphorylation and activation of the RET receptor tyrosine kinase (By similarity). SUBCELLULAR LOCATION: Cell membrane; lipid-anchor; GPI-anchor (By similarity). SIMILARITY: Belongs to the GDNFR family.

Western blot analysis indicates a high level of specificity to GFR alpha-3 for this antiserum.
Application Information:
IHC, WB. A dilution of 1:500 to 1:1000 is recommended for both applications. This antibody stains nicely large neurons in the rat DRG. Specifically, if you would like to perform WB using rat brain homogenate detecting GFR alpha-3, this is the antibody to be used. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: GFR alpha-3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Mouse GFR alpha-3 protein (aa: 1-11)
Host: Rabbit
Isotype: IgG
Reactivity: Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge