Prostaglandin D₂ Express ELISA kit
Lieferant: CAYMAN
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
512041-96 512041-480 512041-96S 512041-480S
CAYM512041-96EA 581 EUR InStock
CAYM512041-96 CAYM512041-480 CAYM512041-96S CAYM512041-480S
Prostaglandin D₂ Express ELISA kit
Tests ELISAs
Prostaglandin D₂ Express ELISA kit is a competitive assay that can be used for quantification of PGD₂.
  • Measure PGD₂ in multiple sample types
  • Assay 24 samples in triplicate or 36 samples in duplicate
  • Measure PGD₂ levels down to 350 pg/ml
  • Get results in under 4 hours
Prostaglandin D₂ (PGD₂) is biosynthesised in the brain by a soluble, 26 kDa glutathione-independent lipocalin-type PGD₂ synthase. PGD₂ accumulates in the cerebrospinal fluid (CSF), where it induces physiologic sleep in rats and humans. PGD₂ is also synthesised in mast cells and leukocytes by a cellular, myeloid-type, glutathione-dependent PGD synthase. This PGD₂ which is formed in the intracellular and vascular compartments is rapidly metabolised to 11β-PGF2α and other metabolites.
Measurement of the parent eicosanoid PGD₂ is appropriate in cell culture lysates and in CSF, where concentrations of several hundred pg/ml have been measured. PGD₂ Express ELISA Kit is a competitive assay that can be used for quantification of PGD₂ in cell culture lysates and enzymatic reactions. This ELISA offers the convenience of a fast assay with a two hour incubation time and one hour development time, while still achieving an assay range of approximately 117,2 to 15000 pg/ml and a sensitvity (80% B/B₀) of approximately 350 pg/ml.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge
Diese Webseite verwendet Cookies, eigene oder von Drittanbieter, zur Sammlung statistischer Informationen, um Inhalte und Anzeigen für Sie zu personalisieren. Wenn Sie diese Seite weiterhin verwenden, akzeptieren Sie die Cookie-Nutzung.
Weitere Informationen finden Sie in unseren Datenschutzbestimmungen..