Immersion probe
Lieferant: COMARK
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
PX33L
COMAPX33LEA 153 EUR InStock
COMAPX33L
Immersion probe
Temperaturfühler
The PX33L Integral Plug Probe is a general purpose penetration probe that connects directly to a thermometer for single-handed temperature taking.
  • Temperature range: −40 to 150 °C / −40 to 302 °F
  • Sensor type: Thermistor (PST)
  • Thin tip: 3,3 mm
The PX33L Integral Plug Probe is fitted with a 6-Pin Lumberg connector with locking screw, to provide a strong and secure probe-to-instrument connection with better prevention of liquid ingress.
Not suitable for intrinsically safe applications
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge