D014-00EA
32.25
EUR
InStock
D014-00
D014-04
D012-00
D012-02
D013-00
D013-02
D013-04
D014-02
D015-00
D015-02
D016-00
D016-02
D016-04
J.T.Baker® DILUT-IT™, Potassium Phosphate, Dissolution Media Concentrate
Puffer
Biologische Puffer
DILUT-IT is a ready-to-use dissolution media concentrate, which is pre-measured and only need to be diluted with purified water.
DILUT-IT™ dissolution media concentrates are produced from raw materials in accordance with USP/EP/BP/JP guidelines under validated processes in Avantor Performance Materials' ISO certified manufacturing facilities