Cleanroom sleeve protectors, Tyvek® IsoClean®, Model IC501B WS
Lieferant: DuPont
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
IC501BWHWS
DUPOIC501BWHWSEA 201 EUR InStock
DUPOIC501BWHWS
Cleanroom sleeve protectors, Tyvek® IsoClean®, Model IC501B WS
Einweg-Kleidung
These sleeves, model IC501BWH WS, are made of Tyvek®, which is a fabric composed of flash spun high density polyethylene, providing an ideal balance of protection, durability, and comfort. Tyvek® is permeable to both air and water vapour, yet repels water-based liquids and aerosols. It provides an inherent barrier to particles, microorganisms, and non-hazardous light liquid splash.
  • Ultra-low linting fabric
  • Bound internal seems
  • Tunnelled elastic at wrist and bicep
  • 450 mm length
  • Aseptically folded
  • Gamma irradiated to Sterility Assurance Level (SAL) of 10⁻⁶
  • Suitable for use in GMP class A/B (ISO Class 5) cleanrooms
All DuPont™ Tyvek® IsoClean® sterile accessories (option code WS) are packed in a dual barrier packaging system, consisting of an inner and outer easy-tear cleanroom bag.
Garments and accessories made of sterile Tyvek® IsoClean® are typically used in cleanrooms within the hospital pharmacies, biotech, pharmaceutical, medical device manufacturing, food processing, cosmetics industry, as well as in other critical or controlled environments.
Zertifizierungen: Partial body chemical protective clothing, Type PB [6-B], EN 14126, Cat. III
Verpackung: 100 per box, individually packed in pairs. Subgrouped by 25 in an outer bag. Two polyethylene liners. Cardboard box.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge