Drucken
Revolve R4 Microscope EX-Demo Model
Lieferant: DISCOVER ECHO
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
RVSF1000-01450
ECHLRVSF1000-01450EA 38900 EUR
ECHLRVSF1000-01450
Revolve R4 Microscope EX-Demo Model
Mikroskope
Intuitive touch interface, measurement annotations, length, area, cell count, scale bar, Z-stack.
  • 5 MP CMOS monochrome camera (fluorescence)
  • 12 MP colour camera (brightfield)
  • 12,9" liquid Retina® touch display
  • Output: USB, ethernet, cloud based
LED 50,000 hour lifespan, 5 imaging channels ,4 epi-fluorescence and one transmitted light.