TAE buffer, 50X
Lieferant: EDVOTEK
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
607
EDVO607EA 33 EUR InStock
EDVO607
TAE buffer, 50X
Puffer Elektrophorese-Puffer Laufpuffer
This 50X concentrated solution (Tris-acetate, EDTA, pH 7,8) is sufficient for making 5 L of diluted working buffer for making agarose gels and electrophoresis chamber buffer.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge